Assay Details
| Gene Symbol: ERG28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100877.3 | NP_563858.1 | 2 - 3 | 545 | 73 | |
| GenBank mRNA | AK227737.1 | - | 2 - 3 | 416 | 73 | |
| BT008773.1 | - | 2 - 3 | 326 | 73 | ||
More Information
Set Membership: |
|

ERG28
Genomic Map