Assay Details
| Gene Symbol: ECA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100887.3 | NP_563860.1 | 26 - 27 | 2541 | 76 | |
| GenBank mRNA | AK229199.1 | - | 26 - 27 | 2541 | 76 | |
| EU082212.1 | - | 26 - 27 | 2352 | 76 | ||
More Information
Set Membership: |
|

ECA3
Genomic Map