Assay Details
| Gene Symbol: emb2004 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100925.2 | NP_563871.1 | 3 - 4 | 670 | 128 | |
| GenBank mRNA | AY056236.1 | - | 3 - 4 | 666 | 128 | |
| AY091215.1 | - | 3 - 4 | 534 | 128 | ||
More Information
Set Membership: |
|

emb2004
Genomic Map