Assay Details
| Gene Symbol: NAT8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001331912.1 | NP_001320848.1 | 8 - 9 | 1120 | 72 | |
| NM_100929.5 | NP_172524.1 | 8 - 9 | 1203 | 72 | ||
| GenBank mRNA | AY063935.1 | - | 8 - 9 | 950 | 72 | |
| AY096464.1 | - | 8 - 9 | 931 | 72 | ||
More Information
Set Membership: |
|

NAT8
Genomic Map