Assay Details
| Gene Symbol: NAT8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001331912.1 | NP_001320848.1 | 9 - 10 | 1195 | 71 | |
| NM_100929.5 | NP_172524.1 | 9 - 10 | 1278 | 71 | ||
| GenBank mRNA | AY063935.1 | - | 9 - 10 | 1025 | 71 | |
| AY096464.1 | - | 9 - 10 | 1006 | 71 | ||
More Information
Set Membership: |
|

NAT8
Genomic Map