Assay Details
| Gene Symbol: PHS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084039.1 | NP_001077508.1 | 7 - 8 | 1026 | 119 | |
| NM_001331923.1 | NP_001318974.1 | 7 - 8 | 867 | 119 | ||
| GenBank mRNA | DQ132645.1 | - | 7 - 8 | 1026 | 119 | |
More Information
Set Membership: |
|

PHS1
Genomic Map