Assay Details
| Gene Symbol: Z-ISO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123792.1 | NP_001117264.1 | 1 - 2 | 423 | 150 | |
| NM_001331932.1 | NP_001323336.1 | 1 - 2 | 534 | 150 | ||
| NM_100959.5 | NP_563879.1 | 1 - 2 | 541 | 150 | ||
| GenBank mRNA | AF348578.1 | - | 1 - 2 | 410 | 150 | |
| AK221585.1 | - | 1 - 2 | 420 | 150 | ||
More Information
Set Membership: |
|

Z-ISO
Genomic Map