Assay Details
| Gene Symbol: MLO4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001331947.1 | NP_001320915.1 | 10 - 11 | 1059 | 125 | |
| NM_001331948.1 | NP_001320916.1 | 11 - 12 | 1051 | 125 | ||
| NM_100975.3 | NP_563882.1 | 10 - 11 | 1067 | 125 | ||
| GenBank mRNA | AF369565.1 | - | 10 - 11 | 974 | 125 | |
More Information
Set Membership: |
|

MLO4
Genomic Map