Assay Details
| Gene Symbol: MLO4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001331947.1 | NP_001320915.1 | 12 - 13 | 1276 | 108 | |
| NM_001331948.1 | NP_001320916.1 | 13 - 14 | 1268 | 108 | ||
| NM_100975.3 | NP_563882.1 | 12 - 13 | 1284 | 108 | ||
| GenBank mRNA | AF369565.1 | - | 12 - 13 | 1191 | 108 | |
More Information
Set Membership: |
|

MLO4
Genomic Map