Assay Details
| Gene Symbol: MLO4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001331947.1 | NP_001320915.1 | 6 - 7 | 809 | 83 | |
| NM_001331948.1 | NP_001320916.1 | 7 - 8 | 801 | 83 | ||
| NM_100975.3 | NP_563882.1 | 6 - 7 | 817 | 83 | ||
| GenBank mRNA | AF369565.1 | - | 6 - 7 | 724 | 83 | |
More Information
Set Membership: |
|

MLO4
Genomic Map