Assay Details
Gene Symbol: MLO4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331947.1 | NP_001320915.1 | 7 - 8 | 889 | 90 | |
NM_001331948.1 | NP_001320916.1 | 8 - 9 | 881 | 90 | ||
NM_100975.3 | NP_563882.1 | 7 - 8 | 897 | 90 | ||
GenBank mRNA | AF369565.1 | - | 7 - 8 | 804 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
