Assay Details
| Gene Symbol: AT1G11100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198029.1 | NP_001184958.1 | 1 - 2 | 142 | 69 | |
| NM_001331954.1 | NP_001320182.1 | 1 - 2 | 426 | 69 | ||
| NM_001331955.1 | NP_001320184.1 | 1 - 2 | 426 | 69 | ||
| NM_100983.5 | NP_172577.2 | 1 - 2 | 200 | 69 | ||
| GenBank mRNA | AB493447.1 | - | 1 - 2 | 142 | 69 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001331954.1 transcripts.
Set Membership: |
|

AT1G11100
Genomic Map