Assay Details
| Gene Symbol: AT1G11100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001331954.1 | NP_001320182.1 | 2 - 3 | 502 | 64 | |
| NM_001331955.1 | NP_001320184.1 | 2 - 3 | 502 | 64 | ||
| NM_100983.5 | NP_172577.2 | 2 - 3 | 276 | 64 | ||
| GenBank mRNA | AB493447.1 | - | 2 - 3 | 218 | 64 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001331954.1 transcripts.
Set Membership: |
|

AT1G11100
Genomic Map