Assay Details
| Gene Symbol: AT1G11100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198029.1 | NP_001184958.1 | 3 - 4 | 441 | 128 | |
| NM_001331955.1 | NP_001320184.1 | 3 - 4 | 728 | 128 | ||
| NM_100983.5 | NP_172577.2 | 3 - 4 | 502 | 128 | ||
| GenBank mRNA | AB493447.1 | - | 3 - 4 | 444 | 128 | |
More Information
Set Membership: |
|

AT1G11100
Genomic Map