Assay Details
| Gene Symbol: AT1G11100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001331956.1 | NP_001320183.1 | 5 - 6 | 1656 | 97 | |
| NM_100983.5 | NP_172577.2 | 7 - 8 | 2133 | 97 | ||
More Information
Set Membership: |
|

AT1G11100
Genomic Map