Assay Details
| Gene Symbol: AT1G11100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198029.1 | NP_001184958.1 | 7 - 8 | 2558 | 130 | |
| NM_001331954.1 | NP_001320182.1 | 7 - 8 | 2873 | 130 | ||
| NM_001331955.1 | NP_001320184.1 | 7 - 8 | 2845 | 130 | ||
| NM_001331956.1 | NP_001320183.1 | 6 - 7 | 2010 | 130 | ||
| NM_100983.5 | NP_172577.2 | 8 - 9 | 2487 | 130 | ||
| GenBank mRNA | AB493447.1 | - | 7 - 8 | 2561 | 130 | |
More Information
Set Membership: |
|

AT1G11100
Genomic Map