Assay Details
| Gene Symbol: SUB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001331960.1 | NP_001321706.1 | 11 - 12 | 1771 | 86 | |
| NM_100986.4 | NP_172580.2 | 12 - 13 | 2255 | 86 | ||
| GenBank mRNA | AF399923.1 | - | 12 - 13 | 2044 | 86 | |
| AK227100.1 | - | 12 - 13 | 2056 | 86 | ||
| AY091019.1 | - | 12 - 13 | 2106 | 86 | ||
| AY117318.1 | - | 12 - 13 | 1828 | 86 | ||
| FJ708632.1 | - | 12 - 13 | 1828 | 86 | ||
More Information
Set Membership: |
|

SUB
Genomic Map