Assay Details
| Gene Symbol: SUB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001331960.1 | NP_001321706.1 | 13 - 14 | 2037 | 69 | |
| NM_100986.4 | NP_172580.2 | 14 - 15 | 2521 | 69 | ||
| GenBank mRNA | AF399923.1 | - | 14 - 15 | 2310 | 69 | |
| AY091019.1 | - | 14 - 15 | 2372 | 69 | ||
| AY117318.1 | - | 14 - 15 | 2094 | 69 | ||
| FJ708632.1 | - | 14 - 15 | 2094 | 69 | ||
More Information
Set Membership: |
|

SUB
Genomic Map