Assay Details
| Gene Symbol: SUB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001331960.1 | NP_001321706.1 | 1 - 2 | 163 | 75 | |
| NM_100986.4 | NP_172580.2 | 2 - 3 | 647 | 75 | ||
| GenBank mRNA | AF399923.1 | - | 2 - 3 | 436 | 75 | |
| AK227100.1 | - | 2 - 3 | 448 | 75 | ||
| AY091019.1 | - | 2 - 3 | 498 | 75 | ||
| AY117318.1 | - | 2 - 3 | 220 | 75 | ||
| FJ708632.1 | - | 2 - 3 | 220 | 75 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001331960.1 transcripts.
Set Membership: |
|

SUB
Genomic Map