Assay Details
| Gene Symbol: SUB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001331960.1 | NP_001321706.1 | 6 - 7 | 519 | 74 | |
| NM_100986.4 | NP_172580.2 | 7 - 8 | 1003 | 74 | ||
| GenBank mRNA | AF399923.1 | - | 7 - 8 | 792 | 74 | |
| AK227100.1 | - | 7 - 8 | 804 | 74 | ||
| AY091019.1 | - | 7 - 8 | 854 | 74 | ||
| AY117318.1 | - | 7 - 8 | 576 | 74 | ||
| FJ708632.1 | - | 7 - 8 | 576 | 74 | ||
More Information
Set Membership: |
|

SUB
Genomic Map