Assay Details
| Gene Symbol: SYP125 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100997.2 | NP_172591.1 | 1 - 2 | 663 | 89 | |
| GenBank mRNA | DQ056451.1 | - | 1 - 2 | 460 | 89 | |
More Information
Set Membership: |
|

SYP125
Genomic Map