Assay Details
| Gene Symbol: GAMT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125013.3 | NP_200441.2 | 1 - 2 | 575 | 81 | |
| GenBank mRNA | BT015738.1 | - | 1 - 2 | 543 | 81 | |
| BT020177.1 | - | 1 - 2 | 511 | 81 | ||
More Information
Set Membership: |
|

GAMT2
Genomic Map