Assay Details
| Gene Symbol: ACA8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001345196.1 | NP_001332277.1 | 6 - 7 | 1079 | 54 | |
| NM_125016.2 | NP_200444.1 | 6 - 7 | 962 | 54 | ||
More Information
Set Membership: |
|

ACA8
Genomic Map