Assay Details
| Gene Symbol: AT5G56400 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125023.2 | NP_200451.2 | 2 - 3 | 1078 | 91 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT5G56400| Gene Symbol: AT5G56400 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125023.2 | NP_200451.2 | 2 - 3 | 1078 | 91 | |
Set Membership: |
|