Assay Details
| Gene Symbol: Cpn60beta3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037006.4 | NP_001032083.1 | 14 - 15 | 2037 | 73 | |
| NM_125033.5 | NP_200461.4 | 14 - 15 | 1998 | 73 | ||
| GenBank mRNA | AK319025.1 | - | 14 - 15 | 1876 | 73 | |
More Information
Set Membership: |
|

Cpn60beta3
Genomic Map