Assay Details
| Gene Symbol: PRF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125043.4 | NP_200471.3 | 2 - 3 | 259 | 68 | |
| GenBank mRNA | AY084285.1 | - | 2 - 3 | 355 | 68 | |
| U43592.1 | - | 2 - 3 | 300 | 68 | ||
More Information
Set Membership: |
|

PRF3
Genomic Map