Assay Details
| Gene Symbol: MIOX5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001345210.1 | NP_001332424.1 | 2 - 3 | 299 | 126 | |
| NM_125047.3 | NP_200475.1 | 3 - 4 | 324 | 126 | ||
More Information
Set Membership: |
|

MIOX5
Genomic Map