Assay Details
| Gene Symbol: MIOX5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001345210.1 | NP_001332424.1 | 8 - 9 | 883 | 79 | |
| NM_125047.3 | NP_200475.1 | 9 - 10 | 908 | 79 | ||
More Information
Set Membership: |
|

MIOX5
Genomic Map