Assay Details
| Gene Symbol: BGAL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125070.3 | NP_200498.1 | 1 - 2 | 553 | 71 | |
| GenBank mRNA | AJ270300.1 | - | 1 - 2 | 209 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

BGAL4| Gene Symbol: BGAL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125070.3 | NP_200498.1 | 1 - 2 | 553 | 71 | |
| GenBank mRNA | AJ270300.1 | - | 1 - 2 | 209 | 71 | |
Set Membership: |
|