Assay Details
| Gene Symbol: BGAL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125070.3 | NP_200498.1 | 6 - 7 | 1056 | 99 | |
| GenBank mRNA | AJ270300.1 | - | 6 - 7 | 712 | 99 | |
More Information
Set Membership: |
|

BGAL4
Genomic Map