Assay Details
| Gene Symbol: AT5G56880 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125071.3 | NP_200499.1 | 1 | 552 | 70 | |
| Gene Symbol: BGAL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125070.3 | NP_200498.1 | 17 | 3298 | 70 | |
More Information
Set Membership: |
|

AT5G56880
Genomic Map