Assay Details
| Gene Symbol: EMB1789 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125075.3 | NP_200503.1 | 5 - 6 | 1224 | 94 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

EMB1789| Gene Symbol: EMB1789 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125075.3 | NP_200503.1 | 5 - 6 | 1224 | 94 | |
Set Membership: |
|