Assay Details
| Gene Symbol: LUT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125085.5 | NP_200513.1 | 5 - 6 | 1091 | 128 | |
| GenBank mRNA | AY040024.1 | - | 5 - 6 | 918 | 128 | |
| AY079371.1 | - | 5 - 6 | 813 | 128 | ||
More Information
Set Membership: | 
		   
					
						
							 | 
				

LUT2
Genomic Map