Assay Details
| Gene Symbol: LUT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125085.5 | NP_200513.1 | 6 - 7 | 1236 | 80 | |
| GenBank mRNA | AY040024.1 | - | 6 - 7 | 1063 | 80 | |
| AY079371.1 | - | 6 - 7 | 958 | 80 | ||
More Information
Set Membership: |
|

LUT2
Genomic Map