Assay Details
| Gene Symbol: EIR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001345238.1 | NP_001331451.1 | 2 - 3 | 848 | 101 | |
| NM_125091.4 | NP_568848.1 | 2 - 3 | 822 | 101 | ||
| GenBank mRNA | AF056026.1 | - | 2 - 3 | 591 | 101 | |
| AF086906.1 | - | 2 - 3 | 646 | 101 | ||
| AF093241.1 | - | 2 - 3 | 596 | 101 | ||
| AK176358.1 | - | 2 - 3 | 629 | 101 | ||
| AY078965.1 | - | 2 - 3 | 625 | 101 | ||
| BT001152.1 | - | 2 - 3 | 573 | 101 | ||
More Information
Set Membership: |
|

EIR1
Genomic Map