Assay Details
| Gene Symbol: EIR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001345238.1 | NP_001331451.1 | 5 - 6 | 1830 | 70 | |
| NM_125091.4 | NP_568848.1 | 5 - 6 | 1804 | 70 | ||
| GenBank mRNA | AF056026.1 | - | 5 - 6 | 1573 | 70 | |
| AF086906.1 | - | 5 - 6 | 1628 | 70 | ||
| AF093241.1 | - | 5 - 6 | 1578 | 70 | ||
| AK176358.1 | - | 5 - 6 | 1611 | 70 | ||
| AY078965.1 | - | 5 - 6 | 1607 | 70 | ||
| BT001152.1 | - | 5 - 6 | 1555 | 70 | ||
More Information
Set Membership: |
|

EIR1
Genomic Map