Assay Details
| Gene Symbol: AT5G57130 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125095.4 | NP_568849.2 | 2 - 3 | 1786 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT5G57130| Gene Symbol: AT5G57130 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125095.4 | NP_568849.2 | 2 - 3 | 1786 | 69 | |
Set Membership: |
|