Assay Details
| Gene Symbol: ATLIG4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001345245.1 | NP_001332576.1 | 2 - 3 | 186 | 70 | |
| NM_125098.4 | NP_568851.2 | 11 - 12 | 1310 | 70 | ||
| GenBank mRNA | AF233527.1 | - | 11 - 12 | 1230 | 70 | |
More Information
Set Membership: |
|

ATLIG4
Genomic Map