Assay Details
| Gene Symbol: ATLIG4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001345245.1 | NP_001332576.1 | 8 - 9 | 708 | 144 | |
| NM_125098.4 | NP_568851.2 | 17 - 18 | 1832 | 144 | ||
| GenBank mRNA | AF233527.1 | - | 17 - 18 | 1752 | 144 | |
More Information
Set Membership: |
|

ATLIG4
Genomic Map