Assay Details
| Gene Symbol: GS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125126.3 | NP_568858.1 | 3 - 4 | 484 | 137 | |
| GenBank mRNA | AY063967.1 | - | 3 - 4 | 392 | 137 | |
| AY084531.1 | - | 3 - 4 | 392 | 137 | ||
| AY114031.1 | - | 3 - 4 | 363 | 137 | ||
More Information
Set Membership: |
|

GS1
Genomic Map