Assay Details
| Gene Symbol: ZFP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125133.5 | NP_200560.1 | 1 | 681 | 100 | |
| GenBank mRNA | BT024687.1 | - | 1 | 314 | 100 | |
| L39645.1 | - | 1 | 377 | 100 | ||
More Information
Set Membership: |
|

ZFP2
Genomic Map