Assay Details
| Gene Symbol: BIO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125140.4 | NP_200567.2 | 6 - 7 | 1546 | 93 | |
| GenBank mRNA | EU089963.1 | - | 6 - 7 | 1541 | 93 | |
| HQ857557.1 | - | 6 - 7 | 1482 | 93 | ||
More Information
Set Membership: |
|

BIO1
Genomic Map