Assay Details
| Gene Symbol: BIO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125140.4 | NP_200567.2 | 7 - 8 | 1637 | 66 | |
| GenBank mRNA | EU089963.1 | - | 7 - 8 | 1632 | 66 | |
| HQ857557.1 | - | 7 - 8 | 1573 | 66 | ||
More Information
Set Membership: |
|

BIO1
Genomic Map