Assay Details
| Gene Symbol: AT5G57770 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001345271.1 | NP_001332540.1 | 1 - 2 | 224 | 67 | |
| NM_125160.6 | NP_200585.3 | 1 - 2 | 244 | 67 | ||
More Information
Set Membership: |
|

AT5G57770
Genomic Map