Assay Details
| Gene Symbol: CER3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125164.3 | NP_200588.2 | 1 - 2 | 453 | 62 | |
| GenBank mRNA | AB099512.1 | - | 1 - 2 | 425 | 62 | |
| AB116548.1 | - | 1 - 2 | 134 | 62 | ||
| AY131334.1 | - | 1 - 2 | 424 | 62 | ||
More Information
Set Membership: |
|

CER3
Genomic Map