Assay Details
| Gene Symbol: CER3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125164.3 | NP_200588.2 | 2 - 3 | 685 | 128 | |
| GenBank mRNA | AB099512.1 | - | 2 - 3 | 657 | 128 | |
| AB116548.1 | - | 2 - 3 | 366 | 128 | ||
| AY131334.1 | - | 2 - 3 | 656 | 128 | ||
More Information
Set Membership: |
|

CER3
Genomic Map