Assay Details
| Gene Symbol: CER3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125164.3 | NP_200588.2 | 8 - 9 | 1847 | 76 | |
| GenBank mRNA | AB099512.1 | - | 8 - 9 | 1819 | 76 | |
| AB116548.1 | - | 8 - 9 | 1528 | 76 | ||
| AY131334.1 | - | 8 - 9 | 1818 | 76 | ||
More Information
Set Membership: |
|

CER3
Genomic Map