Assay Details
| Gene Symbol: ADCL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001345276.1 | NP_001330897.1 | 2 - 3 | 907 | 79 | |
| NM_125170.5 | NP_200593.2 | 4 - 5 | 1081 | 79 | ||
| GenBank mRNA | AK226473.1 | - | 4 - 5 | 1061 | 79 | |
| AY099783.1 | - | 4 - 5 | 1060 | 79 | ||
| AY128874.1 | - | 4 - 5 | 1021 | 79 | ||
More Information
Set Membership: |
|

ADCL
Genomic Map