Assay Details
| Gene Symbol: SKIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125175.2 | NP_568870.1 | 1 - 2 | 347 | 81 | |
| GenBank mRNA | AF263377.1 | - | 1 - 2 | 347 | 81 | |
| AK229464.1 | - | 1 - 2 | 350 | 81 | ||
| BT026067.1 | - | 1 - 2 | 329 | 81 | ||
More Information
Set Membership: |
|

SKIP1
Genomic Map