Assay Details
| Gene Symbol: FIP1[V] | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001345286.1 | NP_001332116.1 | 2 - 3 | 1111 | 87 | |
| NM_125189.3 | NP_200612.2 | 2 - 3 | 1108 | 87 | ||
More Information
Set Membership: |
|

FIP1[V]
Genomic Map